Manganese(IV) oxide, Puratronic™, 99.996% (metals basis)
Manganese(IV) oxide, Puratronic™, 99.996% (metals basis)
Manganese(IV) oxide, Puratronic™, 99.996% (metals basis)
Manganese(IV) oxide, Puratronic™, 99.996% (metals basis)
Thermo Scientific Chemicals

Manganese(IV) oxide, Puratronic™, 99.996% (metals basis)

CAS: 1313-13-9 | MnO2 | 86.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 010805.22
also known as 010805-22
Price (USD)
590.65
Online Exclusive
656.00
Save 65.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
590.65
Online Exclusive
656.00
Save 65.35 (10%)
Each
Chemical Identifiers
CAS1313-13-9
IUPAC Namedioxomanganese
Molecular FormulaMnO2
InChI KeyNUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILESO=[Mn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormPowder
Total Metal Impurities≤0.004% (40 ppm)
It is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only