Ammonium molybdate (para) hydrate, Puratronic™, 99.999% (metals basis excluding W), W 300ppm max
Ammonium molybdate (para) hydrate, Puratronic™, 99.999% (metals basis excluding W), W 300ppm max
Ammonium molybdate (para) hydrate, Puratronic™, 99.999% (metals basis excluding W), W 300ppm max
Ammonium molybdate (para) hydrate, Puratronic™, 99.999% (metals basis excluding W), W 300ppm max
Thermo Scientific Chemicals

Ammonium molybdate (para) hydrate, Puratronic™, 99.999% (metals basis excluding W), W 300ppm max

CAS: 2127839-79-4 | H24Mo7N6O24 | 1163.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 010811.06
also known as 010811-06
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.20
Save 9.55 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.20
Save 9.55 (15%)
Each
Chemical Identifiers
CAS2127839-79-4
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It is used for ceramics, decorating, electroplating, fixing of metals, fertilizers, photography, and as a negative stain in electron microscopy. It is also used in the manufacture of molybdenum. It is a useful reagent for detecting and determining phosphates, arsenates, lead and some alkaloids. It is useful in the preparation of catalysts for desulfurization and dehydrogenation reactions. Ammonium molybdate has been reported to control polycondensation reactions and decrease shrinkage values of metakaolin-based geopolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only