Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max
Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max
Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max
Thermo Scientific Chemicals

Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max

CAS: 1313-27-5 | MoO3 | 143.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010812.14
also known as 010812-14
Price (USD)
85.65
Special offer
Online exclusive
Ends: 28-Dec-2026
100.00
Save 14.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
Ends: 28-Dec-2026
100.00
Save 14.35
Each
Chemical Identifiers
CAS1313-27-5
IUPAC Nametrioxomolybdenum
Molecular FormulaMoO3
InChI KeyJKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILESO=[Mo](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Impurity contentW 300ppm max.
Its chief application is as an oxidation catalyst and as a raw material for the production of molybdenum metal. It is a Precursor to LAMOX fast ion conductors and superconductors. Molybdenum trioxide has also been suggested as a potential anti-microbial agent, e.g., in polymers. In contact with water, it forms H+ ions that can kill bacteria effectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only