Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis)
Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis)
Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis)
Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis)

CAS: 69098-15-3 | Cl2Ni | 129.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 010815.14
also known as 010815-14
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS69098-15-3
IUPAC Namenickel(2+) dichloride
Molecular FormulaCl2Ni
InChI KeyQMMRZOWCJAIUJA-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
It is used in electroplating and as a catalyst for organic conversions, for example in chemo-selective thioacetalization of aldehydes. In combination with lithium aluminum hydride, it serves as a reducing agent for alkenes, alkynes, and organic halides; it can cleave N-O bond and open epoxides. It is a precursor to several nickel-phosphine complexes, such as bis(triphenylphosphine)nickel(II) chloride, which are used in alkyne trimerizations, carbonylations, and as catalysts in organic reactions such as Suzuki-Miyaura cross coupling reactions as an alternative to palladium(0) catalysts. It is the precursor to acetylacetonate complex of Ni, used for producing 1,5-cyclooctadiene complex, an important reagent in organonickel chemistry. It can be used to prepare the sandwich compound nickelocene through dimethoxyethane complex of nickel chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only