Nickel(II) sulfate hydrate, Puratronic™, 99.9985% (metals basis)
Nickel(II) sulfate hydrate, Puratronic™, 99.9985% (metals basis)
Nickel(II) sulfate hydrate, Puratronic™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Nickel(II) sulfate hydrate, Puratronic™, 99.9985% (metals basis)

CAS: 15244-37-8 | NiO4S | 154.749 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010820.14
also known as 010820-14
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS15244-37-8
IUPAC Namenickel(2+) sulfate
Molecular FormulaNiO4S
InChI KeyLGQLOGILCSXPEA-UHFFFAOYSA-L
SMILES[Ni++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0015% max.
Nickel(II) sulfate hydrate is used in electroplating and as a chemical intermediate to produce other nickel compounds. It is also used in printing and in dyeing of textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only