Selenous acid, Puratronic™, 99.999% (metals basis)
Selenous acid, Puratronic™, 99.999% (metals basis)
Selenous acid, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Selenous acid, Puratronic™, 99.999% (metals basis)

CAS: 7783-00-8 | H2O3Se | 128.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010855.14
also known as 010855-14
Price (USD)
310.65
Online Exclusive
345.00
Save 34.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
310.65
Online Exclusive
345.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS7783-00-8
IUPAC Nameselenous acid
Molecular FormulaH2O3Se
InChI KeyMCAHWIHFGHIESP-UHFFFAOYSA-N
SMILESO[Se](O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Selenous acid is used in steel guns, wherein it changes the color of the steel from silver-grey to blue-grey. It serves as an important component of the Mecke reagent, which is utilized for drug testing. It acts as a source of selenium. It acts as a reagent for alkaloids and an oxidizing agent. Further, it is used in labeling radiopharmaceuticals. It is involved in the organic synthesis for the preparation of glyoxal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only