Sodium carbonate, anhydrous, Puratronic™, 99.997% (metals basis)
Sodium carbonate, anhydrous, Puratronic™, 99.997% (metals basis)
Sodium carbonate, anhydrous, Puratronic™, 99.997% (metals basis)
Thermo Scientific Chemicals

Sodium carbonate, anhydrous, Puratronic™, 99.997% (metals basis)

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 010861.06
also known as 010861-06
Price (USD)
59.65
Online Exclusive
66.30
Save 6.65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
59.65
Online Exclusive
66.30
Save 6.65 (10%)
Each
Chemical Identifiers
CAS497-19-8
IUPAC Namedisodium carbonate
Molecular FormulaCNa2O3
InChI KeyCDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (special)?2.5 % of Theoretical value (Na)
Total Metal Impurities≤0.003% (30 ppm)
FormPowder
Sodium carbonate is commonly used as an inorganic base and a buffer component in several synthetic organic transformations in both chemical industries and research laboratories. Often used as an electrolyte, sodium carbonate acts as a very good conductor in the process of electrolysis. Apart from being used as a common chemical/biological/biotechnological laboratory reagent, it is also used as a water softener in laundering, processing wood pulp, cleaners, and making glass. It can be used for standardization of acids, and qualitative testing of cations. In dyeing with fiber-reactive dyes, sodium carbonate is used to ensure proper chemical bonding of the dye with cellulose fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only