Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis)
Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis)
Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals

Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis)

m.p. approx. 50°C, begins to lose CO2 approx.100°C, converts to Na2CO3 | CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 010863.09
also known as 010863-09
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Sodium hydrogen carbonate is used in a process for removing paint and corrosion called sodablasting; the process is particularly suitable for cleaning aluminum panels which can be distorted by other types of abrasive. It can be administered to pools, spas, and garden ponds to raise pH levels. Also can be used as a fire extinguisher. Its primary use is in cooking (baking), as a leavening agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only