Sodium iodide hydrate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Sodium iodide hydrate, Puratronic™, 99.999% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 010871.22
also known as 010871-22
Price (USD)
360.65
Online Exclusive
400.00
Save 39.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
360.65
Online Exclusive
400.00
Save 39.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
Chemical Name or MaterialSodium iodide hydrate
DOT InformationHazard Class: 9; Packaging Group: III
EINECS Number231-679-3
Formula Weight149.89 (Anhydrous)
View more
Sodium Iodide is used in the synthesis of dye-sensitized solar cells. Also used in the preparation of propargylamines as inhibitors of lysine specific. Demethylase. Used in photography and as analytical reagents. They are used in the measurement of the energy of gamma rays, by measuring the amplitude of pulses of light generated by electrons which are excited by the gamma rays and in the Finkelstein reaction via an SN2 pathway, detection of radiation, and in the conversion of an alkyl chloride into an alkyl iodide. Sodium Iodide is used in PCR and to produce chemokines, particularly CCL2, CXCL8, and CXC14 by stimulation of thyroid follicular cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only