Sodium sulfate, Puratronic™, 99.9955% (metals basis)
Sodium sulfate, Puratronic™, 99.9955% (metals basis)
Sodium sulfate, Puratronic™, 99.9955% (metals basis)
Thermo Scientific Chemicals

Sodium sulfate, Puratronic™, 99.9955% (metals basis)

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010874.14
also known as 010874-14
Price (USD)
206.65
Special offer
Online exclusive
Ends: 30-Jun-2026
243.00
Save 36.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
206.65
Special offer
Online exclusive
Ends: 30-Jun-2026
243.00
Save 36.35 (15%)
Each
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0045% max.
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only