Strontium nitrate, Puratronic™, 99.9965% (metals basis)
Strontium nitrate, Puratronic™, 99.9965% (metals basis)
Strontium nitrate, Puratronic™, 99.9965% (metals basis)
Thermo Scientific Chemicals

Strontium nitrate, Puratronic™, 99.9965% (metals basis)

CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010879.22
also known as 010879-22
Price (USD)
434.65
Online Exclusive
483.00
Save 48.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
434.65
Online Exclusive
483.00
Save 48.35 (10%)
Each
Chemical Identifiers
CAS10042-76-9
IUPAC Namestrontium(2+) dinitrate
Molecular FormulaN2O6Sr
InChI KeyDHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.
Strontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. It mixed with glycolic acid and used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only