Titanium(IV) oxide, Puratronic™, 99.995% (metals basis)
Titanium(IV) oxide, Puratronic™, 99.995% (metals basis)
Titanium(IV) oxide, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Titanium(IV) oxide, Puratronic™, 99.995% (metals basis)

CAS: 13463-67-7 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 010897.09
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS13463-67-7
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Total Metal Impurities≤0.0055% (≤55ppm)
Titanium(IV) oxide is used in paints, varnishes, paper, plastics printing inks, sun screen, fibers, rubber, cosmetic products, anti-fogging coatings, foodstuffs, pure titanium, glass, glass ceramics, memristor, electrical ceramics, catalysts, electric conductors and chemical intermediates. It exhibit excellent photocatalytic properties (Honda-Fujishima effect) In addition, titanium dioxide is also used to cleave protein at the site where proline is present.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only