Vanadium(V) oxide, 99.99% (metals basis)
Vanadium(V) oxide, 99.99% (metals basis)
Vanadium(V) oxide, 99.99% (metals basis)
Thermo Scientific Chemicals

Vanadium(V) oxide, 99.99% (metals basis)

CAS: 1314-62-1 | O5V2 | 181.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 010904.18
also known as 010904-18
Price (USD)
609.65
Special offer
Online exclusive
Ends: 28-Dec-2026
717.00
Save 107.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
609.65
Special offer
Online exclusive
Ends: 28-Dec-2026
717.00
Save 107.35
Each
Chemical Identifiers
CAS1314-62-1
IUPAC Name[(dioxovanadio)oxy]dioxovanadium
Molecular FormulaO5V2
InChI KeyGNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILESO=[V](=O)O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst for the manufacture of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only