Cesium carbonate, 99.9% (metals basis)
Cesium carbonate, 99.9% (metals basis)
Cesium carbonate, 99.9% (metals basis)
Cesium carbonate, 99.9% (metals basis)
Thermo Scientific Chemicals

Cesium carbonate, 99.9% (metals basis)

CAS: 534-17-8 | CCs2O3 | 325.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 010924.22
also known as 010924-22
Price (USD)
170.65
Special offer
Online exclusive
Ends: 30-Jun-2026
201.00
Save 30.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
170.65
Special offer
Online exclusive
Ends: 30-Jun-2026
201.00
Save 30.35 (15%)
Each
Chemical Identifiers
CAS534-17-8
IUPAC Namedicaesium(1+) carbonate
Molecular FormulaCCs2O3
InChI KeyFJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES[Cs+].[Cs+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.

Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only