Nickel wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis)
Nickel wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis)
Nickel wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Nickel wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
5 x 100 cm
Catalog number 010929.BY
also known as 010929-BY
Price (USD)
221.65
Special offer
Online exclusive
Ends: 30-Jun-2026
260.00
Save 38.35 (15%)
Each
Quantity:
100 cm
Request bulk or custom format
Price (USD)
221.65
Special offer
Online exclusive
Ends: 30-Jun-2026
260.00
Save 38.35 (15%)
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.995% (metals basis)
Assay from Supplier's CofA99.995% (metals basis)
FormWire, 1.0mm (0.04in) diameter, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only