Nickel wire, 0.05mm (0.002in) dia, Puratronic™, 99.99% (met als basis)
Nickel wire, 0.05mm (0.002in) dia, Puratronic™, 99.99% (met als basis)
Nickel wire, 0.05mm (0.002in) dia, Puratronic™, 99.99% (met als basis)
Thermo Scientific Chemicals

Nickel wire, 0.05mm (0.002in) dia, Puratronic™, 99.99% (met als basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
25 m
Catalog number 010933.G5
also known as 010933-G5
Price (USD)
74.60
Each
Quantity:
5 m
Request bulk or custom format
Price (USD)
74.60
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace MetalTotal metal impurities less than 0.01%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only