Iron wire, 0.25mm (0.01in) dia, Puratronic™, 99.99% (metals basis)
Iron wire, 0.25mm (0.01in) dia, Puratronic™, 99.99% (metals basis)
Iron wire, 0.25mm (0.01in) dia, Puratronic™, 99.99% (metals basis)
Thermo Scientific Chemicals

Iron wire, 0.25mm (0.01in) dia, Puratronic™, 99.99% (metals basis)

Form: approx. 0.39g/m | CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
25 m
Catalog number 010937.G9
also known as 010937-G9
Price (USD)
1,090.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,283.00
Save 192.35 (15%)
Each
Quantity:
25 m
Request bulk or custom format
Price (USD)
1,090.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,283.00
Save 192.35 (15%)
Each
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.25 mm (0.01 in.) diameter
Total Metal Impurities≤0.01%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only