Cobalt wire, 0.25mm (0.01in) dia, Puratronic™, 99.995% (metals basis)
Cobalt wire, 0.25mm (0.01in) dia, Puratronic™, 99.995% (metals basis)
Cobalt wire, 0.25mm (0.01in) dia, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Cobalt wire, 0.25mm (0.01in) dia, Puratronic™, 99.995% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 010948.G1
also known as 010948-G1
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Quantity:
1 m
Request bulk or custom format
Price (USD)
150.65
Special offer
Online exclusive
Ends: 28-Dec-2026
177.00
Save 26.35
Each
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue silver
FormWire, 0.25mm (0.01in) diameter.
Assay from Supplier's CofA≥99.995% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only