Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)

Form: approx. 1.1g/25x200mm | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 200 mm
25 x 1000 mm
Catalog number 010950.HV
also known as 010950-HV
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.20
Save 14.55 (15%)
Each
Quantity:
25 x 200 mm
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.20
Save 14.55 (15%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Copper
Total Metal Impurities0.001% max.
FormFoil
Comment0.023 - 0.028mm thick

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only