Gold wire, 1.0mm (0.04in) dia, Premion™, 99.9985% (metals basis)
Gold wire, 1.0mm (0.04in) dia, Premion™, 99.9985% (metals basis)
Gold wire, 1.0mm (0.04in) dia, Premion™, 99.9985% (metals basis)
Gold wire, 1.0mm (0.04in) dia, Premion™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Gold wire, 1.0mm (0.04in) dia, Premion™, 99.9985% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 cm
10 cm
50 cm
Catalog number 010965.BP
also known as 010965-BP
Price (USD)
92.70
Each
Quantity:
2 cm
Request bulk or custom format
Price (USD)
92.70
Each
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.0mm (0.04in) diameter
Appearance (Color)Golden
Assay from Supplier's CofA99.9985% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only