Potassium iodide, 99.995% (metals basis)
Potassium iodide, 99.995% (metals basis)
Potassium iodide, 99.995% (metals basis)
Thermo Scientific Chemicals

Potassium iodide, 99.995% (metals basis)

CAS: 7681-11-0 | IK | 166.003 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 010978.14
also known as 010978-14
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
161.00
Save 24.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
161.00
Save 24.35 (15%)
Each
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
Potassium iodide is used with silver nitrate to make silver iodide an important chemical in film photography. Medicinally it is used for thyroid blockade, thyroid storm and also as an expectorant. It is a component in some hair treatment chemicals. It is also used as a fluorescence quenching agent in biomedical research, an application that takes advantage of collisional quenching of fluorescent substances by the iodide ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only