Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals
Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals
Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010980.04
Price (USD)
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS7789-23-3
IUPAC Namepotassium fluoride
Molecular FormulaFK
InChI KeyNROKBHXJSPEDAR-UHFFFAOYSA-M
SMILES[F-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals
Assay (special)?2.5 % of Theoretical value (K)
Total Metal Impurities≤0.01% (100 ppm)
Potassium fluoride is used in organic chemistry as a fluorinating agent to replace other halide ions like chorine, bromine etc. It is used as a catalyst in the manufacture of polyurethanes. Further, it is widely used for etching of glass, in disinfectant formulations and manufacture of optical glass. It is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only