Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln.
Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln.
Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln.
Thermo Scientific Chemicals

Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln.

Air Sensitive | CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Catalog number 010991.AA
also known as 010991-AA
Price (USD)
47.65
Online exclusive
52.60
Save 4.95
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
47.65
Online exclusive
52.60
Save 4.95
Each
Chemical Identifiers
CAS10035-10-6
IUPAC Namehydrogen bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-N
SMILESBr
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤ 0.0001%
Appearance (Color)Colorless to yellow
FormLiquid
Concentration (weight/weight)44-49%
Hydrobromic acid is used for the production of inorganic bromides of zinc, calcium, and sodium. It is used as a catalyst in some alkylation reactions. In the field of metallurgy, it is employed in the extraction of certain ores. Further, it is used as a reagent for the cleavage of certain ether linkages or as a brominating agent in organic transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only