Cesium iodide, 99.999% (metals basis)
Cesium iodide, 99.999% (metals basis)
Cesium iodide, 99.999% (metals basis)
Thermo Scientific Chemicals

Cesium iodide, 99.999% (metals basis)

CAS: 7789-17-5 | CsI | 259.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010992.14
also known as 010992-14
Price (USD)
132.65
Online exclusive
147.00
Save 14.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
132.65
Online exclusive
147.00
Save 14.35
Each
Chemical Identifiers
CAS7789-17-5
IUPAC Namecaesium(1+) iodide
Molecular FormulaCsI
InChI KeyXQPRBTXUXXVTKB-UHFFFAOYSA-M
SMILES[I-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Cesium iodide is used as an input phosphor in x-ray image intensifier tube of fluoroscopy equipment. It is also used as an equipment for electromagnetic calorimetry in experimental particle physics. It is often employed as a beamsplitter in Fourier Transform Infrared (FT-IR) spectrometers. Cesium iodide is an inorganic scintillator which is mainly used in experimental nuclear physics, high energy physics, nuclear medicine, nuclear tomography, environmental studies and many other fields of use. It is utilized in calorimeters and detectors and also employed as a phosphor in fluoroscopy equipments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only