Calcium carbonate, 99.95% (metals basis)
Calcium carbonate, 99.95% (metals basis)
Calcium carbonate, 99.95% (metals basis)
Thermo Scientific Chemicals

Calcium carbonate, 99.95% (metals basis)

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010996.22
also known as 010996-22
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Calcium carbonate is used in the purification of iron, refining of sugar from sugar beet, and flue gas desulfurization applications in fossil fuel power stations. It is used in blackboard chalk, printing and writing paper, paints, plastics, PVC cables, polypropylene compounds, thermosetting resins, adhesives, sealants, and decorating fillers. It is an abrasive, and an ingredient in many glazes. It is also used in the pharmaceutical industry as an inert filler for tablets and other pharmaceuticals. It is used as a base in many organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only