Tin powder, -100 mesh, 99.995% (metals basis)
Tin powder, -100 mesh, 99.995% (metals basis)
Tin powder, -100 mesh, 99.995% (metals basis)
Thermo Scientific Chemicals

Tin powder, -100 mesh, 99.995% (metals basis)

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
250 g
Catalog number 011013.04
also known as 011013-04
Price (USD)
37.65
Online exclusive
41.80
Save 4.15
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
37.65
Online exclusive
41.80
Save 4.15
Each
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only