Indium powder, -325 mesh, 99.99% (metals basis)
Indium powder, -325 mesh, 99.99% (metals basis)
Indium powder, -325 mesh, 99.99% (metals basis)
Thermo Scientific Chemicals

Indium powder, -325 mesh, 99.99% (metals basis)

CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 011024.06
also known as 011024-06
Price (USD)
127.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
127.00
Each
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.01%
FormPowder, -325 mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only