Rhodium(III) iodide, 99.9% (metals basis), Rh 20.8% min
Rhodium(III) iodide, 99.9% (metals basis), Rh 20.8% min
Rhodium(III) iodide, 99.9% (metals basis), Rh 20.8% min
Thermo Scientific Chemicals

Rhodium(III) iodide, 99.9% (metals basis), Rh 20.8% min

CAS: 15492-38-3 | I3Rh | 483.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number 011029.03
also known as 011029-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS15492-38-3
IUPAC Namerhodium(3+) triiodide
Molecular FormulaI3Rh
InChI KeyKXAHUXSHRWNTOD-UHFFFAOYSA-K
SMILES[Rh+3].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Rhodium(III) iodide is applied for carbonylation process. It is also used as a precursor for hydroformylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only