Platinum(II) chloride, 99.9% (metals basis), Pt 73% min
Platinum(II) chloride, 99.9% (metals basis), Pt 73% min
Platinum(II) chloride, 99.9% (metals basis), Pt 73% min
Thermo Scientific Chemicals

Platinum(II) chloride, 99.9% (metals basis), Pt 73% min

CAS: 10025-65-7 | Cl2Pt | 265.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 011044.06
also known as 011044-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS10025-65-7
IUPAC Nameplatinum(2+) dichloride
Molecular FormulaCl2Pt
InChI KeyCLSUSRZJUQMOHH-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pt++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II). It is a well known chemotherapy drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only