Stainless Steel powder, -100 mesh, Type 304-L
Stainless Steel powder, -100 mesh, Type 304-L
Stainless Steel powder, -100 mesh, Type 304-L
Thermo Scientific Chemicals

Stainless Steel powder, -100 mesh, Type 304-L

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2 kg
Catalog number 011085.18
also known as 011085-18
Price (USD)
21.60
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
21.60
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni; 70:19:11 wt%
Chemical Name or MaterialStainless Steel powder, Type 304-L
Health Hazard 1H317-H350-H372
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
MDL NumberMFCD00148499
View more
Stainless Steel powder, -100 mesh, Type 304-L is the most common stainless steel alloy widely used in wire weaving. It has excellent corrosion resistance and can withstand temperatures of 1100 degrees F. It has better welding characteristics for lower carbon content.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only