Stainless Steel powder, -100 mesh, Type 316-L
Thermo Scientific Chemicals

Stainless Steel powder, -100 mesh, Type 316-L

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 011089.36
also known as 011089-36
Price (USD)
88.10
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
88.10
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Chemical Name or MaterialStainless Steel powder, Type 316-L
ColorGray
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H319-H317-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
View more
It has an extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment. It finds its application in diamond tools, grinding and P/M parts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only