Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis)
Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis)
Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis)
Thermo Scientific Chemicals

Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis)

CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 011116.04
also known as 011116-04
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.50
Save 6.85 (14%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.50
Save 6.85 (14%)
Each
Chemical Identifiers
CAS7778-18-9
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.007% max.
Calcium sulfate, anhydrous is used as adhesives and sealant chemical, Fillers, Fuels and fuel additives, Intermediates, Process regulators, Agricultural Products, Building/Construction Materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only