Barium chromate, Puratronic™, 99.995% (metals basis)
Barium chromate, Puratronic™, 99.995% (metals basis)
Barium chromate, Puratronic™, 99.995% (metals basis)
Barium chromate, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Barium chromate, Puratronic™, 99.995% (metals basis)

CAS: 10294-40-3 | BaCrO4 | 253.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 011124.14
also known as 011124-14
Price (USD)
262.65
Online Exclusive
292.00
Save 29.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
262.65
Online Exclusive
292.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS10294-40-3
IUPAC Namebarium(2+) dioxochromiumbis(olate)
Molecular FormulaBaCrO4
InChI KeyQFFVPLLCYGOFPU-UHFFFAOYSA-N
SMILES[Ba++].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
Barium chromate has been found to be useful in many capacities. often the compound is used as a carrier for the chromium ions. is an oxidizing agent, making it useful as a burn rate modifier in pyrotechnic compositions. It is especially useful in delay compositions such as delay fuses. It is also used in the composition of a catalyst for alkane dehydration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only