Barium acetate, Puratronic™, 99.999% (metals basis)
Barium acetate, Puratronic™, 99.999% (metals basis)
Barium acetate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Barium acetate, Puratronic™, 99.999% (metals basis)

CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011126.14
also known as 011126-14
Price (USD)
248.65
Special offer
Online exclusive
Ends: 28-Dec-2026
292.00
Save 43.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
248.65
Special offer
Online exclusive
Ends: 28-Dec-2026
292.00
Save 43.35
Each
Chemical Identifiers
CAS543-80-6
IUPAC Namebarium(2+) diacetate
Molecular FormulaC4H6BaO4
InChI KeyITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES[Ba++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only