Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO)
Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO)
Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO)

CAS: 13760-80-0 | F3Yb | 230.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number 011189.18
also known as 011189-18
Price (USD)
817.65
Special offer
Online exclusive
Ends: 30-Jun-2026
962.00
Save 144.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
817.65
Special offer
Online exclusive
Ends: 30-Jun-2026
962.00
Save 144.35 (15%)
Each
Chemical Identifiers
CAS13760-80-0
IUPAC Nameytterbium(3+) trifluoride
Molecular FormulaF3Yb
InChI KeyXASAPYQVQBKMIN-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Yb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Total Rare Earth Oxide Impurities≤0.02%
Ytterbium(III) fluoride is used in the preparation and study of fluorinated glasses. Further, it is employed as a laboratory reagent and in optical glasses and structural ceramics. It finds application in catalysts, electrical components and photo-optical materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only