Ytterbium(III) nitrate hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Ytterbium(III) nitrate hydrate, REacton™, 99.99% (REO)

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011196.18
also known as 011196-18
Price (USD)
419.65
Online Exclusive
466.00
Save 46.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
419.65
Online Exclusive
466.00
Save 46.35 (10%)
Each
Specifications
Assay Percent Notes(REO)
Chemical Name or MaterialYtterbium(III) nitrate hydrate
ColorWhite
DOT InformationTransport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECS Number237-384-6
View more
Ytterbium(III) nitrate hydrate, is used as a precursor of nanoscale coatings of carbon composites. Source of yttrium used to make yttrium-based surfactant mesophases which are promising as adsorbing agents as well as for optically functional species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only