Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thulium(III) chloride hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Thulium(III) chloride hydrate, REacton™, 99.9% (REO)

CAS: 19423-86-0 | Cl3Tm | 275.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 011200.09
also known as 011200-09
Price (USD)
357.65
Special offer
Online exclusive
Ends: 30-Jun-2026
420.00
Save 62.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
357.65
Special offer
Online exclusive
Ends: 30-Jun-2026
420.00
Save 62.35 (15%)
Each
Chemical Identifiers
CAS19423-86-0
IUPAC Namethulium(3+) trichloride
Molecular FormulaCl3Tm
InChI KeyILOTUXNTERMOJL-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Tm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Thulium(III) chloride hydrate is used in proteomics research. It is also used in the production of glass, phosphors and lasers. Further, it serves as an important dopant for fiber amplifiers. Further, it is also used to prepare thulium(III) oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only