Terbium(III,IV) oxide, REacton™, 99.99% (REO)
Terbium(III,IV) oxide, REacton™, 99.99% (REO)
Terbium(III,IV) oxide, REacton™, 99.99% (REO)
Terbium(III,IV) oxide, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Terbium(III,IV) oxide, REacton™, 99.99% (REO)

CAS: 12037-01-3 | O7Tb4 | 747.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 011208.14
also known as 011208-14
Price (USD)
851.65
Online Exclusive
946.00
Save 94.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
851.65
Online Exclusive
946.00
Save 94.35 (10%)
Each
Chemical Identifiers
CAS12037-01-3
IUPAC Nametetraoxotetraterboxane
Molecular FormulaO7Tb4
InChI KeyUWZVBPNIJCKCMC-UHFFFAOYSA-N
SMILESO=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Terbium(III,IV) oxide is used in the preparation of other terbium compounds. Activating agents for fluorescent powders, composites for catalytic automobile exhaust converters, Glass materials with a Faraday Rotation effect for optical and laser-based devices, Magneto-optical glasses, Magneto-optical recording materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only