Samarium(III) fluoride, anhydrous, REacton™, 99.99% (REO)
Samarium(III) fluoride, anhydrous, REacton™, 99.99% (REO)
Samarium(III) fluoride, anhydrous, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Samarium(III) fluoride, anhydrous, REacton™, 99.99% (REO)

CAS: 13765-24-7 | F3Sm | 207.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 011226.18
also known as 011226-18
Price (USD)
563.65
Online Exclusive
626.00
Save 62.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
563.65
Online Exclusive
626.00
Save 62.35 (10%)
Each
Chemical Identifiers
CAS13765-24-7
IUPAC Namesamarium(3+) trifluoride
Molecular FormulaF3Sm
InChI KeyOJIKOZJGHCVMDC-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Sm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Samarium(III) fluoride is used as laboratory reagent, in optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. It has specialized uses in phosphors, lasers, and thermoelectric devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only