Samarium(III) oxide, REacton™, 99.9% (REO)
Samarium(III) oxide, REacton™, 99.9% (REO)
Samarium(III) oxide, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Samarium(III) oxide, REacton™, 99.9% (REO)

CAS: 12060-58-1 | Sm2O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
Catalog number 011229.22
also known as 011229-22
Price (USD)
230.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
230.00
Each
Chemical Identifiers
CAS12060-58-1
IUPAC Namedisamarium(3+) trioxidandiide
Molecular FormulaO3Sm2
InChI KeyFKTOIHSPIPYAPE-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sm+3].[Sm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. It finds use as a neutron absorber in nuclear power reactors to control the reaction. It is also used as a catalyst for the dehydration of primary alcohols to aldehydes or ketones. It forms samarium perchlorate when treated with perchloric acid, which is used as a reference material for checking the wavelength scale of a spectrophotometer in the range of 200 to 500 nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only