Samarium(III) chloride hydrate, REacton™, 99.9% (REO)
Samarium(III) chloride hydrate, REacton™, 99.9% (REO)
Samarium(III) chloride hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Samarium(III) chloride hydrate, REacton™, 99.9% (REO)

CAS: 10361-82-7 | Cl3Sm | 256.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011231.14
also known as 011231-14
Price (USD)
77.50
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
77.50
Each
Chemical Identifiers
CAS10361-82-7
IUPAC Namesamarium(3+) trichloride
Molecular FormulaCl3Sm
InChI KeyBHXBZLPMVFUQBQ-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Sm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Samarium(III) chloride hydrate is used for the preparation of samarium metal, which finds use as magnets. It is combined with sodium chloride to get a low melting point eutectic mixture. Further, it serves as a precursor for the preparation of other samarium salts and bis(pentamethylcyclopentadienyl)alkylsamarium(III) complexes. In addition, it serves as a catalyst for hydrogenation and hydrosilylation of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only