Praseodymium(III) chloride hydrate, REacton™, 99.9% (REO)
Praseodymium(III) chloride hydrate, REacton™, 99.9% (REO)
Praseodymium(III) chloride hydrate, REacton™, 99.9% (REO)
Praseodymium(III) chloride hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Praseodymium(III) chloride hydrate, REacton™, 99.9% (REO)

CAS: 19423-77-9 | Cl3Pr | 247.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011237.14
also known as 011237-14
Price (USD)
86.10
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
86.10
Each
Chemical Identifiers
CAS19423-77-9
IUPAC Namepraseodymium(3+) trichloride
Molecular FormulaCl3Pr
InChI KeyLHBNLZDGIPPZLL-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Pr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
FormCrystalline aggregates
Total Rare Earth Oxide Impurities0.1% max.
Aqueous solutions of praseodymium(III) chloride can be used to prepare insoluble praseodymium(III) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only