Neodymium(III) oxide, REacton™, 99.9% (REO)
Neodymium(III) oxide, REacton™, 99.9% (REO)
Neodymium(III) oxide, REacton™, 99.9% (REO)
Neodymium(III) oxide, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Neodymium(III) oxide, REacton™, 99.9% (REO)

CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 011248.18
also known as 011248-18
Price (USD)
112.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
112.00
Each
Chemical Identifiers
CAS1313-97-9
IUPAC Namedineodymium(3+) trioxidandiide
Molecular FormulaNd2O3
InChI KeyPLDDOISOJJCEMH-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Nd+3].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Neodymium(III) oxide is used as a catalyst in polymer chemistry. Glasses doped with neodymium(III) oxide are dichroic in nature and is used in welding goggles and other colored glasses. It finds use in solid-state lasers, carbon arc-light electrodes and enamels. It serves as the starting material for the synthesis of other neodymium(III) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only