Neodymium(III) chloride hydrate, REacton™, 99.99% (REO)
Neodymium(III) chloride hydrate, REacton™, 99.99% (REO)
Neodymium(III) chloride hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Neodymium(III) chloride hydrate, REacton™, 99.99% (REO)

CAS: 19423-78-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011252.22
also known as 011252-22
Price (USD)
338.65
Online Exclusive
376.00
Save 37.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
338.65
Online Exclusive
376.00
Save 37.35 (10%)
Each
Chemical Identifiers
CAS19423-78-0
IUPAC Nameneodymium(3+) trichloride
Molecular FormulaCl3Nd
InChI KeyATINCSYRHURBSP-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only