Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade
Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade
Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade
Thermo Scientific Chemicals

Lanthanum(III) oxide, 99.99% (REO), Ca 10ppm max, AAS Grade

CAS: 1312-81-8 | La2O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 011266.18
also known as 011266-18
Price (USD)
63.65
Online Exclusive
70.90
Save 7.25 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.90
Save 7.25 (10%)
Each
Chemical Identifiers
CAS1312-81-8
IUPAC Namedilanthanum(3+) trioxidandiide
Molecular FormulaLa2O3
InChI KeyMRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[La+3].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Impurity contentCa ≤ 10ppm
Total Rare Earth Oxide Impurities≤0.01% (100 ppm)
FormPowder
Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacturing of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics. In addition, it is used as a catalyst to effect certain organic transformations. It is one of the active ingredients and finds application in thermoelectric and piezoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only