Holmium(III) chloride hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Holmium(III) chloride hydrate, REacton™, 99.99% (REO)

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011276.09
also known as 011276-09
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Specifications
Assay Percent Notes(REO)
Chemical Name or MaterialHolmium(III) chloride hydrate
EINECS Number233-387-1
Formula Weight271.29 (Anhydrous)
Health Hazard 1H315-H319-H335
View more
Holmium(III) chloride is used in nuclear control rods, microwaves, cubic zirconia, coloring glass yellow, solid state lasers and magnetic poles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only