Holmium(III) chloride hexahydrate, REacton™, 99.9% (REO)
Holmium(III) chloride hexahydrate, REacton™, 99.9% (REO)
Holmium(III) chloride hexahydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Holmium(III) chloride hexahydrate, REacton™, 99.9% (REO)

CAS: 14914-84-2 | Cl3H12HoO6 | 379.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011277.14
also known as 011277-14
Price (USD)
141.65
Online Exclusive
157.00
Save 15.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
141.65
Online Exclusive
157.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS14914-84-2
IUPAC Nametrichloroholmium hexahydrate
Molecular FormulaCl3H12HoO6
InChI KeyBUQFCXABQYNXLP-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.Cl[Ho](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Holmium(III) chloride hexahydrate is used as laboratory reagent and also used in proteomics research , optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only