Europium(III) nitrate hydrate, REacton™, 99.99% (REO)
Europium(III) nitrate hydrate, REacton™, 99.99% (REO)
Europium(III) nitrate hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Europium(III) nitrate hydrate, REacton™, 99.99% (REO)

CAS: 100587-95-9 | EuN3O9 | 337.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 011296.06
also known as 011296-06
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS100587-95-9
IUPAC Nameeuropium(3+) trinitrate
Molecular FormulaEuN3O9
InChI KeyGAGGCOKRLXYWIV-UHFFFAOYSA-N
SMILES[Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Europium(III) nitrate hydrate is widely used as dopant and used in the preparation of europium-doped hafnium and europium doped gadolinium oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only