Europium(III) chloride hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Europium(III) chloride hydrate, REacton™, 99.9% (REO)

EuCl3·xH2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 011298.06
also known as 011298-06
Price (USD)
114.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
114.00
Each
Specifications
Assay Percent Range99.9% (REO)
Chemical Name or MaterialEuropium(III) chloride hydrate
ColorYellow
EINECS Number233-040-4
Formula Weight258.32 (anhydrous)
View more
Europium(III) chloride hydrate is used as a catalyst in organic synthesis. It is involved in the preparation of amino acid based europium(III) complex in order to prepare photoactive thin films, which finds application in organic light emitting diodes. It is also used as a precursor for the preparation of other europium compounds and its salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only