Dysprosium(III) chloride hydrate, 99.99% (REO)
Thermo Scientific Chemicals

Dysprosium(III) chloride hydrate, 99.99% (REO)

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 011317.06
also known as 011317-06
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.70
Save 7.05 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.70
Save 7.05 (15%)
Each
Chemical Identifiers
CAS10025-74-8
IUPAC Namedysprosium(3+) trichloride
Molecular FormulaCl3Dy
InChI KeyBOXVSFHSLKQLNZ-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Dy+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
FormPowder or crystalline powder
CommentDysprosium oxide: 47.0-49.0%
Total Rare Earth Oxide Impurities≤0.01% (≤100ppm)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only